Gene Transcripts

Gene Symbol
Flnc
Entrez Gene ID
362332
Gene Name
filamin C, gamma
Gene Aliases
ABP-L, FLN-C
Chromosome Location
Chr. 4: 56710930 - 56738779 on Build Rnor_6.0
UniGene ID
Rn.22352
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191862.1 NP_001178791.1 23 4181
XM_006236214.2 XP_006236276.1 23 4337

View Full Product Details