Gene Transcripts

Gene Symbol
FCGBP
Entrez Gene ID
8857
Gene Name
Fc fragment of IgG binding protein
Gene Aliases
FC(GAMMA)BP
Chromosome Location
Chr. 19: 39863323 - 39934626 on Build GRCh38
UniGene ID
Hs.111732
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003890.2 NP_003881.2 14703
GenBank mRNA BM781647.1 - 1 374
D84239.1 - 14703

View Full Product Details