Gene Transcripts

Gene Symbol
Ap3s2
Entrez Gene ID
683402
Gene Name
adaptor-related protein complex 3, sigma 2 subunit
Gene Aliases
Chromosome Location
Chr. 1: 141612961 - 141655417 on Build Rnor_6.0
UniGene ID
Rn.3618
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001115039.1 NP_001108511.1 2 295
GenBank mRNA BC158881.1 - 2 295

View Full Product Details