Gene Transcripts

Gene Symbol
Habp4
Entrez Gene ID
56541
Gene Name
hyaluronic acid binding protein 4
Gene Aliases
4933413D03Rik, 4933428J01Rik
Chromosome Location
Chr. 13: 64161866 - 64186537 on Build GRCm38
UniGene ID
Mm.40989
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019986.3 NP_064370.2 2 623
GenBank mRNA AF227684.1 - 2 566
AK016800.1 - 2 666
AK144206.1 - 2 607
AK146525.1 - 2 596
BC082806.1 - 2 625
BC119460.2 - 1 120
BC148304.2 - 1 127

View Full Product Details