Gene Transcripts

Gene Symbol
ITM2A
Entrez Gene ID
9452
Gene Name
integral membrane protein 2A
Gene Aliases
BRICD2A, E25A
Chromosome Location
Chr. X: 79360384 - 79367552 on Build GRCh38
UniGene ID
Hs.17109
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004867.4 NP_004858.1 2, 3 585
GenBank mRNA AF038953.1 - 2, 3 387
AK223327.1 - 2, 3 364
AK313154.1 - 2, 3 543
AY359051.1 - 2, 3 326
BC040437.1 - 2, 3 340 5748519
BG506587.1 - 2, 3 584
CR456875.1 - 2, 3 248

View Full Product Details