Gene Transcripts

Gene Symbol
GMEB2
Entrez Gene ID
26205
Gene Name
glucocorticoid modulatory element binding protein 2
Gene Aliases
GMEB-2, P79PIF, PIF79
Chromosome Location
Chr. 20: 63587594 - 63627101 on Build GRCh38
UniGene ID
Hs.473286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012384.4 NP_036516.1 8, 9 1030
XM_005260202.4 XP_005260259.1 8, 9 1241
XM_006723776.2 XP_006723839.1 6, 7 682
XM_011528779.2 XP_011527081.1 6, 7 746
GenBank mRNA AB033095.1 - 8, 9 907
AF173867.1 - 7, 8 1306
AL133646.1 - 6, 7 754
BC036305.1 - 8, 9 957 5166938
BQ954281.1 - 7, 8 732

View Full Product Details