Gene Transcripts

Gene Symbol
TEDDM1
Entrez Gene ID
127670
Gene Name
transmembrane epididymal protein 1
Gene Aliases
EDDM9, Epdd1, HE9, HEL-S-45e, TMEM45C
Chromosome Location
Chr. 1: 182398117 - 182400616 on Build GRCh38
UniGene ID
Hs.156977
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172000.3 NP_741997.3 1 851
GenBank mRNA AJ515384.2 - 1 851
GQ472227.1 - 1 851

View Full Product Details