Gene Transcripts
- Gene Symbol
- DEUP1
- Entrez Gene ID
- 159989
- Gene Name
- deuterosome assembly protein 1
- Gene Aliases
- CCDC67
- Chromosome Location
- Chr. 11: 93329079 - 93438470 on Build GRCh38
- UniGene ID
- Hs.436625
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_181645.3 |
NP_857596.2
|
9, 10 |
1138 |
|
XM_005273802.1 |
XP_005273859.1
|
9, 10 |
1185 |
|
XM_011542629.2 |
XP_011540931.1
|
9, 10 |
1168 |
|
XM_011542630.1 |
XP_011540932.1
|
9, 10 |
1185 |
|
XM_011542631.2 |
XP_011540933.1
|
9, 10 |
2245 |
|
XM_011542632.2 |
XP_011540934.1
|
9, 10 |
1168 |
|
XM_011542633.2 |
XP_011540935.1
|
9, 10 |
1168 |
|
XM_011542635.2 |
XP_011540937.1
|
9, 10 |
1168 |
|
XM_011542636.2 |
XP_011540938.1
|
3, 4 |
512 |
|
XM_011542638.2 |
XP_011540940.1
|
3, 4 |
508 |
|
XM_017017245.1 |
XP_016872734.1
|
9, 10 |
1168 |
|
GenBank mRNA |
AK058122.1 |
- |
7, 8 |
872 |
|
AK127973.1 |
- |
9, 10 |
1185 |
|
BC017929.1 |
- |
9, 10 |
1117 |
|
BC031247.1 |
- |
3, 4 |
392 |
5268521 |
View Full Product Details