Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144606.5 | NP_653207.1 | 4 | 562 | |
NM_144997.5 | NP_659434.2 | 4 | 562 | ||
XM_011523714.2 | XP_011522016.1 | 4 | 559 | ||
XM_011523716.1 | XP_011522018.1 | 2 | 937 | ||
XM_011523718.2 | XP_011522020.1 | 3 | 443 | ||
XM_011523719.2 | XP_011522021.1 | 4 | 559 | ||
XM_011523721.2 | XP_011522023.1 | 3 | 308 | ||
XM_017024304.1 | XP_016879793.1 | 5 | 728 | ||
XM_017024305.1 | XP_016879794.1 | 5 | 743 | ||
XM_017024306.1 | XP_016879795.1 | 5 | 786 | ||
XM_017024307.1 | XP_016879796.1 | 5 | 748 | ||
XM_017024308.1 | XP_016879797.1 | 6 | 924 | ||
XM_017024309.1 | XP_016879798.1 | 4 | 559 | ||
GenBank mRNA | AF517523.1 | - | 4 | 513 | |
AK126951.1 | - | 1 | 1399 | ||
AK127912.1 | - | 4 | 562 | ||
AK309336.1 | - | 4 | 559 | ||
BC015687.2 | - | 4 | 513 | 4684974 | |
BQ423946.1 | - | 567 | |||
CX759825.1 | - | 4 | 525 | ||
KU178888.1 | - | 1 | 58 | ||
KU178889.1 | - | 1 | 58 |