Gene Transcripts

Gene Symbol
ASTL
Entrez Gene ID
431705
Gene Name
astacin-like metallo-endopeptidase (M12 family)
Gene Aliases
SAS1B
Chromosome Location
Chr. 2: 96122817 - 96143059 on Build GRCh38
UniGene ID
Hs.447993
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002036.3 NP_001002036.3 5 425
XM_011511205.2 XP_011509507.1 4 1240
XM_011511206.2 XP_011509508.1 5 1463
XM_011511207.2 XP_011509509.1 4 1241
XM_011511208.2 XP_011509510.1 4 1241
GenBank mRNA AJ537600.2 - 5 425
BC107126.1 - 5 424
BC107127.1 - 5 425 40023980

View Full Product Details