Gene Transcripts
- Gene Symbol
- KCNQ5-IT1
- Entrez Gene ID
- 100507381
- Gene Name
- KCNQ5 intronic transcript 1
- Gene Aliases
- Chromosome Location
- Chr. 6: 72630495 - 72678558 on Build GRCh38
- UniGene ID
- Hs.662412
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_120503.1 |
|
2 |
176 |
|
GenBank mRNA |
BX104605.1 |
- |
2 |
176 |
|
HY037383.1 |
- |
2 |
317 |
|
View Full Product Details