Gene Transcripts

Gene Symbol
FCGBP
Entrez Gene ID
8857
Gene Name
Fc fragment of IgG binding protein
Gene Aliases
FC(GAMMA)BP
Chromosome Location
Chr. 19: 39863323 - 39934626 on Build GRCh38
UniGene ID
Hs.111732
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003890.2 NP_003881.2 8177
GenBank mRNA BQ065661.1 - 1 189
D84239.1 - 8177

View Full Product Details