Gene Transcripts

Gene Symbol
Gabpb1
Entrez Gene ID
14391
Gene Name
GA repeat binding protein, beta 1
Gene Aliases
BABPB2, E4TF1, E4TF1-47, E4TF1-53, E4Tf1B, GABPB, GABPB-1, GABPB-2, GABPB1-1, GABPB1-2, NRF2B1, NRF2B2
Chromosome Location
Chr. 2: 126628907 - 126676677 on Build GRCm38
UniGene ID
Mm.293266
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271467.1 NP_001258396.1 4 489
NM_001271468.1 NP_001258397.1 3 391
NM_001271469.1 NP_001258398.1 3 391
NM_001271470.1 NP_001258399.1 4 556
NM_001271492.1 NP_001258421.1 3 391
NM_010249.2 NP_034379.1 3 391
NM_207669.2 NP_997552.1 3 391
GenBank mRNA AK040722.1 - 3 392
AK052828.1 - 367
AK139710.1 - 4 556
AK140222.1 - 4 442
AK140907.1 - 3 302
AK146373.1 - 3 255
AY282800.1 - 1 43
AY282801.1 - 3 237
AY282802.1 - 1 43
AY282803.1 - 1 43
AY282804.1 - 3 237
AY282805.1 - 1 43
BC013558.1 - 3 282 3592119
CF730595.1 - 3 244
M74516.1 - 3 286
M74517.1 - 3 285

View Full Product Details