Gene Transcripts

Gene Symbol
IFIT2
Entrez Gene ID
3433
Gene Name
interferon induced protein with tetratricopeptide repeats 2
Gene Aliases
G10P2, GARG-39, IFI-54, IFI-54K, IFI54, IFIT-2, ISG-54 K, ISG-54K, ISG54, P54, cig42
Chromosome Location
Chr. 10: 89301949 - 89309276 on Build GRCh38
UniGene ID
Hs.437609
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001547.4 NP_001538.4 2 1106
GenBank mRNA AK300431.1 - 2 1008
AK312831.1 - 2 1055
BC005987.1 - 2 1009
BC032839.2 - 2 1009 4838844
EB387611.1 - 1 482

View Full Product Details