Gene Transcripts
- Gene Symbol
- Plxnc1
- Entrez Gene ID
- 54712
- Gene Name
- plexin C1
- Gene Aliases
- 2510048K12Rik, AW742158, CD232, Vespr
- Chromosome Location
- Chr. 10: 94790866 - 94945156 on Build GRCm38
- UniGene ID
- Mm.256712
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018797.2 |
NP_061267.1
|
14 |
2776 |
|
GenBank mRNA |
AF190578.1 |
- |
14 |
2776 |
|
View Full Product Details