Gene Transcripts

Gene Symbol
Mcm10
Entrez Gene ID
70024
Gene Name
minichromosome maintenance deficient 10 (S. cerevisiae)
Gene Aliases
2410041F14Rik, AU018508, C330019M07Rik, C79164
Chromosome Location
Chr. 2: 4990724 - 5012848 on Build GRCm38
UniGene ID
Mm.23828
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027290.3 NP_081566.2 16 2389
GenBank mRNA AK030780.1 - 16 2334
BC023236.1 - 8 1251
BC039663.1 - 2 238
BC064314.1 - 16 2752
BC094576.1 - 16 2389 6853824
BC120687.1 - 15 2298 40129610
BC120689.1 - 15 2298 40129612

View Full Product Details