Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027290.3 | NP_081566.2 | 15 | 2244 | |
GenBank mRNA | AK030780.1 | - | 15 | 2189 | |
AK088589.1 | - | 1 | 2336 | ||
BC023236.1 | - | 7 | 1106 | ||
BC039663.1 | - | 1 | 93 | ||
BC064314.1 | - | 15 | 2607 | ||
BC094576.1 | - | 15 | 2244 | 6853824 | |
BC120687.1 | - | 14 | 2153 | 40129610 | |
BC120689.1 | - | 14 | 2153 | 40129612 |