Gene Transcripts

Gene Symbol
Tut1
Entrez Gene ID
70044
Gene Name
terminal uridylyl transferase 1, U6 snRNA-specific
Gene Aliases
2700038E08Rik, PAPD2, Rbm21, TUTase6
Chromosome Location
Chr. 19: 8953850 - 8966210 on Build GRCm38
UniGene ID
Mm.165979
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_197993.2 NP_932110.1 9 2516
GenBank mRNA AK012330.1 - 1 209
BC023900.1 - 9 2493 5321265
BC025499.1 - 9 2540 5320398

View Full Product Details