Gene Transcripts

Gene Symbol
Rnf139
Entrez Gene ID
75841
Gene Name
ring finger protein 139
Gene Aliases
4930555P18Rik
Chromosome Location
Chr. 15: 58889229 - 58902390 on Build GRCm38
UniGene ID
Mm.4537
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175226.4 NP_780435.1 2 738
GenBank mRNA AK028400.1 - 2 691
AK029069.1 - 2 741
AK033506.1 - 2 672
BC052901.1 - 2 687 30070643

View Full Product Details