Gene Transcripts
- Gene Symbol
- Selk
- Entrez Gene ID
- 80795
- Gene Name
- selenoprotein K
- Gene Aliases
- 1110001C03Rik, AA673253, HSPC030, Hsp30
- Chromosome Location
- Chr. 14: 29968380 - 29975074 on Build GRCm38
- UniGene ID
- Mm.104491
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019979.2 |
NP_064363.2
|
2, 3 |
240 |
|
GenBank mRNA |
AF170920.1 |
- |
2, 3 |
269 |
|
AK003206.1 |
- |
2, 3 |
168 |
|
AK006674.1 |
- |
2, 3 |
140 |
|
AK007523.1 |
- |
2, 3 |
152 |
|
AK008419.1 |
- |
2, 3 |
176 |
|
BC038049.1 |
- |
2, 3 |
181 |
2645414 |
BC096044.1 |
- |
2, 3 |
122 |
1248280 |
BC096677.1 |
- |
2, 3 |
170 |
874920 |
BY095166.1 |
- |
2, 3 |
240 |
|
- Gene Symbol
- Gm12060
- Entrez Gene ID
- 100134990
- Gene Name
- selenoprotein K pseudogene
- Gene Aliases
- OTTMUSG00000005275
- Chromosome Location
- Chr. 11: 23558058 - 23558842 on Build GRCm38
- UniGene ID
- Mm.247328
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC027278.1 |
- |
1 |
214 |
|
View Full Product Details