Gene Transcripts

Gene Symbol
Zfhx4
Entrez Gene ID
80892
Gene Name
zinc finger homeodomain 4
Gene Aliases
A930021B15, C130041O22Rik, Zfh-4, Zfh4
Chromosome Location
Chr. 3: 5217420 - 5415861 on Build GRCm38
UniGene ID
Mm.41522
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030708.2 NP_109633.2 2 2537
GenBank mRNA AB024499.1 - 1 2161

View Full Product Details