Gene Transcripts

Gene Symbol
CAMK1D
Entrez Gene ID
57118
Gene Name
calcium/calmodulin dependent protein kinase ID
Gene Aliases
CKLiK, CaM-K1, CaMKID
Chromosome Location
Chr. 10: 12349514 - 12835545 on Build GRCh38
UniGene ID
Hs.600547
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020397.3 NP_065130.1 2 464
NM_153498.3 NP_705718.1 2 464
XM_006717481.3 XP_006717544.1 2 728
XM_006717482.3 XP_006717545.1 2 493
XM_006717483.3 XP_006717546.1 2 455
XM_011519591.2 XP_011517893.1 2 901
XM_017016438.1 XP_016871927.1 3 759
XM_017016439.1 XP_016871928.1 4 590
GenBank mRNA AB081726.1 - 2 420
AF286366.1 - 2 275
AK289393.1 - 3 275
BC035745.1 - 2 281 5748490
BU542765.1 - 2 525
HY168807.1 - 2 465

View Full Product Details