Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018398.2 | NP_060868.2 | 8 | 861 | |
XM_005265318.2 | XP_005265375.1 | 8 | 933 | ||
XM_011533946.2 | XP_011532248.1 | 8 | 933 | ||
XM_011533947.2 | XP_011532249.1 | 8 | 933 | ||
XM_011533948.2 | XP_011532250.1 | 8 | 933 | ||
XM_011533949.2 | XP_011532251.1 | 8 | 933 | ||
XM_011533950.2 | XP_011532252.1 | 8 | 933 | ||
XM_011533951.2 | XP_011532253.1 | 8 | 933 | ||
XM_011533952.2 | XP_011532254.1 | 8 | 933 | ||
XM_011533953.2 | XP_011532255.1 | 8 | 933 | ||
XM_017006850.1 | XP_016862339.1 | 8 | 933 | ||
XM_017006851.1 | XP_016862340.1 | 5 | 515 | ||
XM_017006852.1 | XP_016862341.1 | 8 | 933 | ||
GenBank mRNA | AF516696.1 | - | 8 | 861 | |
AJ272213.1 | - | 7 | 743 | ||
AJ272268.1 | - | 7 | 743 | ||
AK299718.1 | - | 7 | 725 | ||
AK307449.1 | - | 6 | 583 | ||
BC137502.1 | - | 9 | 931 | 9021502 | |
BC137505.1 | - | 9 | 931 | 9021505 |