Gene Transcripts

Gene Symbol
CARD10
Entrez Gene ID
29775
Gene Name
caspase recruitment domain family member 10
Gene Aliases
BIMP1, CARMA3
Chromosome Location
Chr. 22: 37490362 - 37519203 on Build GRCh38
UniGene ID
Hs.57973
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014550.3 NP_055365.2 20 3436
GenBank mRNA AK057334.1 - 5 1448
AK074906.1 - 17 2666
AY028896.1 - 20 3473
AY032928.1 - 20 3436

View Full Product Details