Gene Transcripts

Gene Symbol
ADH6
Entrez Gene ID
130
Gene Name
alcohol dehydrogenase 6 (class V)
Gene Aliases
ADH-5
Chromosome Location
Chr. 4: 99202638 - 99219246 on Build GRCh38
UniGene ID
Hs.586161
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000672.3 NP_000663.1 8 1256
GenBank mRNA BC039065.1 - 7 1109

View Full Product Details