Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304448.1 | NP_001291377.1 | 3 | 615 | |
NM_002259.4 | NP_002250.1 | 2 | 372 | ||
NM_007328.3 | NP_015567.1 | 2 | 372 | ||
NM_213657.2 | NP_998822.1 | 3 | 546 | ||
NM_213658.2 | NP_998823.1 | 3 | 546 | ||
GenBank mRNA | AF461812.1 | - | 2 | 348 | |
AK310992.1 | - | 3 | 615 | ||
BC012550.1 | - | 3 | 550 | 4042168 | |
BC053840.1 | - | 3 | 315 | 5484921 | |
DB464226.1 | - | 2 | 372 | ||
X54867.1 | - | 2 | 322 | ||
X54868.1 | - | 2 | 322 |