Gene Transcripts

Gene Symbol
AIFM2
Entrez Gene ID
84883
Gene Name
apoptosis inducing factor, mitochondria associated 2
Gene Aliases
AMID, PRG3
Chromosome Location
Chr. 10: 70112267 - 70132934 on Build GRCh38
UniGene ID
Hs.650680
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198696.1 NP_001185625.1 9 2530
NM_032797.5 NP_116186.1 9 2527
GenBank mRNA AK027403.1 - 9 2406
AK127353.1 - 9 4282
BX537621.1 - 6 3164

View Full Product Details