Gene Transcripts

Gene Symbol
LINC00612
Entrez Gene ID
253128
Gene Name
long intergenic non-protein coding RNA 612
Gene Aliases
C12orf33
Chromosome Location
Chr. 12: 9055589 - 9065070 on Build GRCh38
UniGene ID
Hs.408739
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_034140.1 2 1313
GenBank mRNA AK123808.1 - 3 1235
BC028195.1 - 2 1313 5163453

View Full Product Details