Gene Transcripts

Gene Symbol
C19orf68
Entrez Gene ID
374920
Gene Name
chromosome 19 open reading frame 68
Gene Aliases
Chromosome Location
Chr. 19: 48170662 - 48197621 on Build GRCh38
UniGene ID
Hs.664054
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199341.3 NP_955373.3 4 3237
XM_005259449.3 XP_005259506.1 4 3675
XM_006723204.3 XP_006723267.1 4 3372
XM_006723205.2 XP_006723268.1 4 3263
XM_011526936.2 XP_011525238.1 3 3277
XM_011526937.2 XP_011525239.1 2 2779
GenBank mRNA BC043386.1 - 5 1926

View Full Product Details