Gene Transcripts
- Gene Symbol
- AZI2
- Entrez Gene ID
- 64343
- Gene Name
- 5-azacytidine induced 2
- Gene Aliases
- AZ2, NAP1, TILP
- Chromosome Location
- Chr. 3: 28322353 - 28349129 on Build GRCh38
- UniGene ID
- Hs.706676
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001134432.1 |
NP_001127904.1
|
2, 3 |
756 |
|
NM_001134433.1 |
NP_001127905.1
|
2, 3 |
756 |
|
NM_001271650.1 |
NP_001258579.1
|
3, 4 |
820 |
|
NM_022461.4 |
NP_071906.1
|
2, 3 |
756 |
|
XM_005265392.2 |
XP_005265449.1
|
3, 4 |
843 |
|
XM_011534009.2 |
XP_011532311.1
|
2, 3 |
296 |
|
XM_011534010.2 |
XP_011532312.1
|
2, 3 |
254 |
|
XM_011534011.2 |
XP_011532313.1
|
2, 3 |
253 |
|
GenBank mRNA |
AK025592.1 |
- |
2, 3 |
603 |
|
AK290637.1 |
- |
2, 3 |
475 |
|
AY151386.1 |
- |
1, 2 |
224 |
|
BG777644.1 |
- |
2, 3 |
711 |
|
BM546093.1 |
- |
1, 2 |
92 |
|
BX386357.2 |
- |
2, 3 |
677 |
|
BX648471.1 |
- |
3, 4 |
484 |
|
CB995808.1 |
- |
2, 3 |
284 |
|
View Full Product Details