Gene Transcripts

Gene Symbol
TMEM243
Entrez Gene ID
79161
Gene Name
transmembrane protein 243
Gene Aliases
C7orf23, MM-TRAG, MMTRAG
Chromosome Location
Chr. 7: 87195593 - 87220603 on Build GRCh38
UniGene ID
Hs.619593
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005250589.3 XP_005250646.1 5 995
XM_017012617.1 XP_016868106.1 5 1118
XM_017012618.1 XP_016868107.1 5 862
GenBank mRNA BC002837.2 - 4 779

View Full Product Details