Gene Transcripts

Gene Symbol
ASF1B
Entrez Gene ID
55723
Gene Name
anti-silencing function 1B histone chaperone
Gene Aliases
CIA-II
Chromosome Location
Chr. 19: 14119509 - 14136628 on Build GRCh38
UniGene ID
Hs.26516
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018154.2 NP_060624.1 4 620
GenBank mRNA AB104486.1 - 4 448
AF279307.1 - 4 541
AK001288.1 - 4 579
AK001466.1 - 4 611
AK223080.1 - 4 574
AK302134.1 - 4 530
BC007726.2 - 4 544 4303318
BC010014.2 - 4 554 4304756
BC036521.1 - 4 584 5270216
CR457235.1 - 4 448

View Full Product Details