Gene Transcripts
- Gene Symbol
- ATG16L1
- Entrez Gene ID
- 55054
- Gene Name
- autophagy related 16 like 1
- Gene Aliases
- APG16L, ATG16A, ATG16L, IBD10, WDR30
- Chromosome Location
- Chr. 2: 233251571 - 233295674 on Build GRCh38
- UniGene ID
- Hs.529322
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001190266.1 |
NP_001177195.1
|
6, 7 |
954 |
|
NM_001190267.1 |
NP_001177196.1
|
6, 7 |
952 |
|
NM_017974.3 |
NP_060444.3
|
6, 7 |
956 |
|
NM_030803.6 |
NP_110430.5
|
6, 7 |
956 |
|
NM_198890.2 |
NP_942593.2
|
3, 4 |
524 |
|
XM_005246082.2 |
XP_005246139.1
|
6, 7 |
893 |
|
XM_005246084.2 |
XP_005246141.1
|
3, 4 |
432 |
|
XM_005246086.2 |
XP_005246143.1
|
3, 4 |
432 |
|
XM_006712608.2 |
XP_006712671.1
|
5, 6 |
704 |
|
GenBank mRNA |
AK000897.1 |
- |
5, 6 |
567 |
|
AK001690.1 |
- |
6, 7 |
802 |
|
AK024453.1 |
- |
6, 7 |
826 |
|
AK027854.1 |
- |
5, 6 |
538 |
|
AK123876.1 |
- |
3, 4 |
423 |
|
AK308400.1 |
- |
6, 7 |
856 |
|
AY358182.1 |
- |
6, 7 |
845 |
|
AY398617.1 |
- |
6, 7 |
845 |
|
BC071846.1 |
- |
3, 4 |
362 |
|
BC117337.1 |
- |
6, 7 |
618 |
|
BC143900.1 |
- |
6, 7 |
618 |
|
EF079889.1 |
- |
6, 7 |
699 |
|
EF079890.1 |
- |
3, 4 |
267 |
|
EF560734.1 |
- |
6, 7 |
826 |
|
JQ924062.1 |
- |
6, 7 |
699 |
|
View Full Product Details