Gene Transcripts

Gene Symbol
ASF1A
Entrez Gene ID
25842
Gene Name
anti-silencing function 1A histone chaperone
Gene Aliases
CGI-98, CIA, HSPC146
Chromosome Location
Chr. 6: 118894077 - 118909171 on Build GRCh38
UniGene ID
Hs.292316
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014034.2 NP_054753.1 4 2247
GenBank mRNA AK025738.1 - 4 2104
AL050261.1 - 4 2101

View Full Product Details