Gene Transcripts
- Gene Symbol
- AMOTL2
- Entrez Gene ID
- 51421
- Gene Name
- angiomotin like 2
- Gene Aliases
- LCCP
- Chromosome Location
- Chr. 3: 134355345 - 134375417 on Build GRCh38
- UniGene ID
- Hs.426312
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278683.1 |
NP_001265612.1
|
3, 4 |
1340 |
|
NM_001278685.1 |
NP_001265614.1
|
3, 4 |
1354 |
|
NM_016201.3 |
NP_057285.3
|
3, 4 |
1354 |
|
XM_006713654.2 |
XP_006713717.1
|
3, 4 |
1372 |
|
XM_011512881.1 |
XP_011511183.1
|
3, 4 |
1211 |
|
XM_017006580.1 |
XP_016862069.1
|
3, 4 |
1211 |
|
XM_017006581.1 |
XP_016862070.1
|
3, 4 |
1392 |
|
XM_017006582.1 |
XP_016862071.1
|
3, 4 |
1219 |
|
GenBank mRNA |
AB023206.1 |
- |
3, 4 |
1287 |
|
AF175966.1 |
- |
2, 3 |
421 |
|
AK001343.1 |
- |
3, 4 |
1217 |
|
AK223598.1 |
- |
3, 4 |
1212 |
|
AK291616.1 |
- |
3, 4 |
1298 |
|
AK299270.1 |
- |
3, 4 |
1340 |
|
BC011454.1 |
- |
1, 2 |
320 |
4181482 |
View Full Product Details