Gene Transcripts

Gene Symbol
ANAPC5
Entrez Gene ID
51433
Gene Name
anaphase promoting complex subunit 5
Gene Aliases
APC5
Chromosome Location
Chr. 12: 121308245 - 121354209 on Build GRCh38
UniGene ID
Hs.7101
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001137559.1 NP_001131031.1 17 2229
NM_016237.4 NP_057321.2 17 2490
XM_005253900.3 XP_005253957.1 17 2453
XM_017019423.1 XP_016874912.1 16 2126
XM_017019424.1 XP_016874913.1 16 2087
GenBank mRNA AK025614.1 - 17 2229
AK054779.1 - 7 2066
AK294139.1 - 10 1486
AK301197.1 - 9 1720
BC001081.2 - 17 2423 2823401
BC001950.1 - 17 2423 2823401
BC003095.1 - 12 1692
BC006301.2 - 17 2427 4100632
BC034243.1 - 16 2130
BC045686.1 - 10 2578
BX381618.2 - 2 373
BX537687.1 - 12 2117

View Full Product Details