Gene Transcripts

Gene Symbol
CNOT8
Entrez Gene ID
9337
Gene Name
CCR4-NOT transcription complex subunit 8
Gene Aliases
CAF1, CALIF, Caf1b, POP2, hCAF1
Chromosome Location
Chr. 5: 154857952 - 154876792 on Build GRCh38
UniGene ID
Hs.26703
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301073.1 NP_001288002.1 7 1713
NM_001301074.1 NP_001288003.1 6 1551
NM_001301075.1 NP_001288004.1 6 1567
NM_001301077.1 NP_001288006.1 7 1625
NM_001301080.1 NP_001288009.1 6 1524
NM_001301082.1 NP_001288011.1 6 1237
NM_001301083.1 NP_001288012.1 5 1357
NM_001301086.1 NP_001288015.1 5 1362
NM_004779.5 NP_004770.4 7 1761
XM_005268527.2 XP_005268584.1 7 2053
XM_011537706.1 XP_011536008.1 8 1735
XM_011537708.2 XP_011536010.1 6 1238
XM_017010048.1 XP_016865537.1 8 1620
XM_017010049.1 XP_016865538.1 8 2215
XM_017010050.1 XP_016865539.1 8 2200
XM_017010051.1 XP_016865540.1 8 2250
XM_017010052.1 XP_016865541.1 7 2005
XM_017010053.1 XP_016865542.1 7 2023
XM_017010054.1 XP_016865543.1 6 1544
XM_017010055.1 XP_016865544.1 6 1576
XM_017010056.1 XP_016865545.1 6 1662
XM_017010057.1 XP_016865546.1 8 1778
XM_017010058.1 XP_016865547.1 5 1366
GenBank mRNA AF053318.1 - 7 1635
AF087844.1 - 6 1463
AF180476.1 - 6 1463
AK025358.1 - 7 1682
AK299589.1 - 8 1593
AL122045.1 - 7 1642
BC017366.2 - 7 1572 3506054
BX647124.1 - 5 1104

View Full Product Details