Gene Transcripts
- Gene Symbol
- AP3B2
- Entrez Gene ID
- 8120
- Gene Name
- adaptor related protein complex 3 beta 2 subunit
- Gene Aliases
- NAPTB
- Chromosome Location
- Chr. 15: 82659281 - 82710112 on Build GRCh38
- UniGene ID
- Hs.199593
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278511.1 |
NP_001265440.1
|
15, 16 |
2085 |
|
NM_001278512.1 |
NP_001265441.1
|
17, 18 |
2238 |
|
NM_004644.4 |
NP_004635.2
|
16, 17 |
2181 |
|
XM_017022640.1 |
XP_016878129.1
|
15, 16 |
2313 |
|
GenBank mRNA |
AF022152.1 |
- |
16, 17 |
2010 |
|
AK097827.1 |
- |
8, 9 |
1078 |
|
BC093739.1 |
- |
16, 17 |
2052 |
7939584 |
BC143342.1 |
- |
15, 16 |
1956 |
9051847 |
BC143346.1 |
- |
17, 18 |
2109 |
9051851 |
CR749860.1 |
- |
15, 16 |
2180 |
|
U37673.1 |
- |
16, 17 |
2037 |
|
View Full Product Details