Gene Transcripts

Gene Symbol
ASAP2
Entrez Gene ID
8853
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 2
Gene Aliases
AMAP2, CENTB3, DDEF2, PAG3, PAP, Pap-alpha, SHAG1
Chromosome Location
Chr. 2: 9206765 - 9405683 on Build GRCh38
UniGene ID
Hs.555902
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135191.1 NP_001128663.1 27 4360
NM_003887.2 NP_003878.1 28 4495
XM_006711898.1 XP_006711961.1 29 4257
XM_006711901.3 XP_006711964.1 29 4107
XM_006711902.1 XP_006711965.1 28 4122
XM_011510403.2 XP_011508705.1 29 4278
XM_011510404.2 XP_011508706.1 28 4268
XM_011510405.2 XP_011508707.1 28 4204
XM_011510406.2 XP_011508708.1 30 4211
XM_011510407.2 XP_011508709.1 28 4143
XM_011510409.1 XP_011508711.1 29 4149
GenBank mRNA AB007860.1 - 28 4495
AK091121.1 - 6 1906
AK124900.1 - 4 2653
BC063308.1 - 27 4313 30345039

View Full Product Details