Gene Transcripts

Gene Symbol
TMCO5A
Entrez Gene ID
145942
Gene Name
transmembrane and coiled-coil domains 5A
Gene Aliases
TMCO5
Chromosome Location
Chr. 15: 37934607 - 37997541 on Build GRCh38
UniGene ID
Hs.179646
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152453.3 NP_689666.2 12 1205
XM_005254168.3 XP_005254225.1 12 1186
GenBank mRNA BC029221.2 - 11 1118

View Full Product Details