Gene Transcripts

Gene Symbol
C7
Entrez Gene ID
730
Gene Name
complement component 7
Gene Aliases
Chromosome Location
Chr. 5: 40909497 - 40982939 on Build GRCh38
UniGene ID
Hs.78065
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000587.2 NP_000578.2 7 819
GenBank mRNA AK290349.1 - 7 829
AK312737.1 - 7 753
BC025402.1 - 7 898
BC063851.1 - 7 800 6184213
J03507.1 - 6 705

View Full Product Details