Gene Transcripts

Gene Symbol
AMELY
Entrez Gene ID
266
Gene Name
amelogenin, Y-linked
Gene Aliases
AMGL, AMGY
Chromosome Location
Chr. Y: 6865918 - 6874056 on Build GRCh38
UniGene ID
Hs.1238
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143.1 NP_001134.1 4, 5 207
XM_017030042.1 XP_016885531.1 5, 6 278
GenBank mRNA AY487421.1 - 4, 5 181
BC069138.1 - 4, 5 207 7216997
BC074976.1 - 4, 5 168 30915513
BC074977.1 - 4, 5 168 30915296
M86933.1 - 4, 5 207

View Full Product Details