Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001715.2 | NP_001706.2 | 13 | 2400 | |
XM_011543824.1 | XP_011542126.1 | 13 | 2499 | ||
XM_011543825.2 | XP_011542127.1 | 13 | 2244 | ||
XM_011543827.1 | XP_011542129.1 | 12 | 2375 | ||
XM_017013730.1 | XP_016869219.1 | 12 | 2297 | ||
GenBank mRNA | AL832875.1 | - | 3 | 1759 | |
BC007371.2 | - | 13 | 1963 | 3634568 | |
BC018703.1 | - | 14 | 2025 | ||
BC032413.1 | - | 13 | 2009 | 5216030 | |
S76617.1 | - | 13 | 2405 | ||
Z33998.1 | - | 13 | 2041 |