Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042533.2 | NP_001035998.1 | 5 | 1294 | |
NM_001261829.1 | NP_001248758.1 | 5 | 827 | ||
NM_032778.5 | NP_116167.3 | 5 | 1294 | ||
NM_153182.3 | NP_694822.2 | 5 | 827 | ||
XM_005247838.4 | XP_005247895.1 | 5 | 1044 | ||
GenBank mRNA | AB083189.1 | - | 5 | 930 | |
AB083190.1 | - | 5 | 930 | ||
AB083191.1 | - | 4 | 747 | ||
AK027299.1 | - | 5 | 777 | ||
AY302110.3 | - | 5 | 997 | ||
AY390536.1 | - | 5 | 979 | ||
AY456380.1 | - | 4 | 354 | ||
BC014928.1 | - | 5 | 981 | 3916148 | |
CR627479.1 | - | 4 | 580 | ||
DQ453796.1 | - | 5 | 791 |