Gene Transcripts

Gene Symbol
BRWD3
Entrez Gene ID
254065
Gene Name
bromodomain and WD repeat domain containing 3
Gene Aliases
BRODL, MRX93
Chromosome Location
Chr. X: 80669488 - 80809736 on Build GRCh38
UniGene ID
Hs.147027
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153252.4 NP_694984.4 8 1054
XM_005262113.3 XP_005262170.1 8 1056
XM_017029384.1 XP_016884873.1 3 460
XM_017029385.1 XP_016884874.1 8 1056
GenBank mRNA AY497046.1 - 8 1054
AY497047.1 - 5 986
AY497048.1 - 8 1054
AY497049.1 - 7 1024
AY497050.1 - 7 903
AY497052.1 - 9 1089
AY497053.1 - 7 903
AY497054.1 - 6 804
AY497055.1 - 9 1089
AY497056.1 - 4 835
AY497057.1 - 4 835
AY497058.1 - 4 674
AY497059.1 - 6 1045
AY497060.1 - 3 736
BC111490.1 - 8 1004

View Full Product Details