Gene Transcripts

Gene Symbol
Asz1
Entrez Gene ID
74068
Gene Name
ankyrin repeat, SAM and basic leucine zipper domain containing 1
Gene Aliases
4933400N19Rik, Gasz, ORF3
Chromosome Location
Chr. 6: 18050964 - 18109061 on Build GRCm38
UniGene ID
Mm.73091
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023729.3 NP_076218.3 1, 2 126
GenBank mRNA AF459789.1 - 1, 2 120
BC058859.1 - 1, 2 135 30011651
BG088136.2 - 1, 2 126

View Full Product Details