Gene Transcripts

Gene Symbol
Ahcyl1
Entrez Gene ID
229709
Gene Name
S-adenosylhomocysteine hydrolase-like 1
Gene Aliases
1110034F20Rik, AA409031, AA414901, Ahcy-rs3, DCAL, Irbit
Chromosome Location
Chr. 3: 107663120 - 107696548 on Build GRCm38
UniGene ID
Mm.220328
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145542.3 NP_663517.2 4, 5 754
GenBank mRNA AB092504.1 - 4, 5 483
BC018218.1 - 4, 5 583 4007102

View Full Product Details