Gene Transcripts

Gene Symbol
Arsb
Entrez Gene ID
11881
Gene Name
arylsulfatase B
Gene Aliases
1110007C02Rik, AI480648, As-1, As-1r, As-1s, As-1t, As1, As1-r, As1-s, As1-t, Asr-1, Ast-1
Chromosome Location
Chr. 13: 93771023 - 93943016 on Build GRCm38
UniGene ID
Mm.300178
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009712.3 NP_033842.3 8 3088
GenBank mRNA AK003524.1 - 1 595
BC055876.1 - 2 1067

View Full Product Details