Gene Transcripts
- Gene Symbol
- LOC101929950
- Entrez Gene ID
- 101929950
- Gene Name
- puromycin-sensitive aminopeptidase-like protein
- Gene Aliases
- Chromosome Location
- Chr. 17: 38078159 - 38118364 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- LOC440434
- Entrez Gene ID
- 440434
- Gene Name
- aminopeptidase puromycin sensitive pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 17: 38195740 - 38257249 on Build GRCh38
- UniGene ID
- Hs.443837
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_036750.2 |
|
2, 3 |
436 |
|
- Gene Symbol
- NPEPPS
- Entrez Gene ID
- 9520
- Gene Name
- aminopeptidase puromycin sensitive
- Gene Aliases
- AAP-S, MP100, PSA
- Chromosome Location
- Chr. 17: 47522950 - 47623276 on Build GRCh38
- UniGene ID
- Hs.443837
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006310.3 |
NP_006301.3
|
2, 3 |
562 |
|
XM_011525496.1 |
XP_011523798.1
|
3, 4 |
364 |
|
XM_017025372.1 |
XP_016880861.1
|
2, 3 |
524 |
|
XM_017025373.1 |
XP_016880862.1
|
3, 4 |
364 |
|
GenBank mRNA |
AJ132583.1 |
- |
2, 3 |
292 |
|
AK095877.1 |
- |
3, 4 |
328 |
|
AK096709.1 |
- |
4, 5 |
349 |
|
AK296887.1 |
- |
2, 3 |
313 |
|
AK297107.1 |
- |
1, 2 |
66 |
|
AK303037.1 |
- |
3, 4 |
357 |
|
AK311414.1 |
- |
|
37 |
|
BC065294.1 |
- |
2, 3 |
251 |
6059589 |
DB476543.3 |
- |
2, 3 |
562 |
|
View Full Product Details