Gene Transcripts

Gene Symbol
Calcrl
Entrez Gene ID
25029
Gene Name
calcitonin receptor-like
Gene Aliases
CLR, Crlr, RATCRLR, RNCLR
Chromosome Location
Chr. 3: 71747940 - 71845487 on Build Rnor_6.0
UniGene ID
Rn.11202
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012717.1 NP_036849.1 6 541
XM_006234438.2 XP_006234500.1 6 796
XM_006234439.2 XP_006234501.1 5 634
GenBank mRNA L27487.1 - 6 814
X70658.1 - 6 541

View Full Product Details